Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAF1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563325
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563325 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563325 Supplier Invitrogen™ Supplier No. PA563325
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PKQPFMLQHA SGEHKDGHGK Highest antigen sequence identity to the following orthologs: Mouse - 40%, Rat - 40%.

TAF1L, also named as TAF (II)210, belongs to the TAF1 family. It may act as a functional substitute for TAF1/TAFII250 during male meiosis, when sex chromosomes are transcriptionally silenced.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAF1L
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TAF1L
Gene Accession No. Q8IZX4
Gene Alias TAF(II)210; TAF1 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 210kDa-like; TAF1L; TAF2A2; TATA-box binding protein associated factor 1 like; TBP-associated factor 1-like; TBP-associated factor 210 kDa; TBP-associated factor RNA polymerase 1-like; transcription initiation factor TFIID 210 kDa subunit; Transcription initiation factor TFIID subunit 1-like
Gene Symbols TAF1L
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TAF1L. Recombinant protein control fragment (Product #RP-100035).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 138474
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.