Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAF1Rabbit anti-Human, Mouse, Rat, Polyclonal, Rockland™
SDP

Catalog No. 501991909
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1909 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-1909 Supplier Rockland Immunochemicals Supplier No. 600401995
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Directed against human TAF1 protein. A BLAST analysis was used to suggest cross-reactivity with TAF1 protein from human, mouse and rat based on 100% homology with the immunizing sequence. Reactivity against homologues from other sources is not known.

3-Phosphoglycerate dehydrogenase (PHGDH; EC 1.1.1.95) catalyzes the transition of 3-phosphoglycerate into 3-phosphohydroxypyruvate, which is the first and rate-limiting step in the phosphorylated pathway of serine biosynthesis, using NAD+/NADH as a cofactor.

Specifications

Antigen TAF1
Applications ChIP Assay, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.1 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene TAF1
Gene Accession No. P21675, Q80UV9
Gene Alias AU015687; B430306D02Rik; BA2R; Ccg1; Ccg-1; CCGS; Cell cycle gene 1 protein; cell cycle, G1 phase defect; complementation of cell cycle block, G1-to-S; DYT3; DYT3/TAF1; KAT4; MRXS33; NSCL2; N-TAF1; OF; p250; RGD1562050; TAF(II)250; TAF1; TAF1 RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF1 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 250kDa; TAF1 RNA polymerase II, TATA box binding protein (TBP)-associated factor, neuron specific isoform; TAF2A; TAFII250; TAFII-250; TATA box binding protein associated factor 1; TATA-box binding protein associated factor 1; TBP-associated factor 250 kDa; transcription factor TFIID p250 polypeptide; Transcription initiation factor TFIID 250 kDa subunit; Transcription initiation factor TFIID subunit 1; XDP
Gene Symbols TAF1
Host Species Rabbit
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 270627, 317256, 6872
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.