Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TAF2 Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP121372DL594
Description
TAF2 Polyclonal antibody specifically detects TAF2 in Human samples. It is validated for Western BlotSpecifications
TAF2 | |
Polyclonal | |
Western Blot | |
150 kDa cofactor of initiator function, CIF150TAFII-150, cofactor of initiator function, 150kD subunit, TAF(II)150, TAF2 RNA polymerase II, TATA box binding protein (TBP)-associated factor, TAF2B, TAFII150RNA polymerase II TBP-associated factor subunit B, TATA box binding protein (TBP)-associated factor, RNA polymerase II, B, 150kD, TBP-associated factor 150 kDa, Transcription initiation factor TFIID 150 kDa subunit, transcription initiation factor TFIID subunit 2 | |
The immunogen recognized by this antibody maps to a region between residue 1149 and 1199 of human TBP-associated factor 2 using the numbering given in entry NP_003175.1 (GeneID 6873). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 594 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
6873 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction