Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ TAF2 Overexpression Lysate
SDP

Catalog No. NBL116686 Shop All R&D Systems Products
Modifier l'affichage
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
NBL116686 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. NBL116686 Fournisseur Novus Biologicals™ Code fournisseur NBL116686
Ajouter au panier
Ajouter au panier
The TAF2 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.

Spécifications

Cell Line The lysate was created in HEK293T cells, using plasmid ID RC213159 and based on accession number NM_3184. The protein contains a C-terminal DDK tag.
For Use With (Application) Western Blot
Quantity 0.1 mg
Storage Buffer RIPA buffer
Purity Protein
Test Specificity Homo sapiens TAF2 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 150kDa (TAF2), mRNA.
Content And Storage -80°C. Avoid freeze-thaw cycles.
Protein State Native

For Research Use Only

missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'

missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.