Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540710
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540710 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540710 Supplier Invitrogen™ Supplier No. PA540710
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: RKRNVLELEI KQDYTSPGTQ KYVGPLKVTV QELDGSFNHT LQIEENSLKH Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 85%.

Initiation of transcription by RNA polymerase II requires the activities of more than 70 polypeptides. The protein that coordinates these activities is transcription factor IID (TFIID), which binds to the core promoter to position the polymerase properly, serves as the scaffold for assembly of the remainder of the transcription complex, and acts as a channel for regulatory signals. TFIID is composed of the TATA-binding protein (TBP) and a group of evolutionarily conserved proteins known as TBP-associated factors or TAFs. TAFs may participate in basal transcription, serve as coactivators, function in promoter recognition or modify general transcription factors (GTFs) to facilitate complex assembly and transcription initiation. This gene encodes one of the larger subunits of TFIID that is stably associated with the TFIID complex. It contributes to interactions at and downstream of the transcription initiation site, interactions that help determine transcription complex response to activators.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAF2
Applications ChIP Assay, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TAF2
Gene Accession No. Q6P1X5
Gene Alias 150 kDa cofactor of initiator function; 150kDa; 4732460C16Rik; AI425886; CIF150; cofactor of initiator function, 150kD subunit; fc30f02; fc56a05; MRT40; RNA polymerase II TBP-associated factor subunit B; TAF(II)150; TAF150; TAF2; taf2 protein; TAF2 RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF2 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 150kDa; TAF2 RNA polymerase II, TATA box binding protein-associated factor, 150 kD; TAF2B; Taf2p; TAFII150; TAFII-150; TAFIIB; TATA box binding protein (TBP)-associated factor 150kDa; TATA box binding protein (TBP)-associated factor, RNA polymerase II, B, 150kD; TATA box binding protein associated factor 2; TATA-box binding protein associated factor 2; TBP-associated factor 150 kDa; TBP-associated factor 2; transcription initiation factor TFIID 150 kDa subunit; Transcription initiation factor TFIID subunit 2; transcription initiation factor TFIID subunit 2-like protein; TSM1; TSM-1; wu:fc30f02; wu:fc56a05; YCR042C; YCR42C; YCR724
Gene Symbols TAF2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human TAF2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6873
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.