Learn More
Invitrogen™ TAF3 Polyclonal Antibody

Description
The antibody detects endogenous levels of total TAF3 protein.
Specifications
Specifications
| Antigen | TAF3 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | TAF3 |
| Gene Accession No. | Q5HZG4, Q5VWG9 |
| Gene Alias | 140 kDa TATA box-binding protein-associated factor; 140kDa; 4933439M23Rik; AW539625; fi76b06; MGC25063; mTAFII140; RNA polymerase II transcription factor TAFII140; RNA polymerase II transcription factor, TFIID subunit; TAF(II)140; TAF140; Taf3; TAF3 RNA polymerase II, TATA box binding protein (TBP)-associated facto; TAF3 RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF3 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 140kDa; TAFII140; TAFII-140; TATA box binding protein associated factor 3; TATA-box binding protein associated factor 3; TBP-associated factor 3; Transcription initiation factor TFIID 140 kDa subunit; transcription initiation factor TFIID subunit 3; Unknown (protein for IMAGE:7281647); wu:fi76b06; zgc:136280 |
| Gene Symbols | TAF3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.