Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAF3 Recombinant Rabbit Monoclonal Antibody (JE64-97)
GREENER_CHOICE

Catalog No. PIMA544701
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544701 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544701 Supplier Invitrogen™ Supplier No. MA544701
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

TAF3 Recombinant Monoclonal Antibody for ICC/IF, IHC (P), Western Blot

Transcription factor TFIID is one of the general factors required for accurate and regulated initiation by RNA polymerase II. TFIID is a multimeric protein complex that plays a central role in mediating promoter responses to various activators and repressors. Required in complex with TBPL2 for the differentiation of myoblasts into myocytes. The complex replaces TFIID at specific promoters at an early stage in the differentiation process. Belongs to the TFIID complex which is composed of TATA binding protein (TBP) and a number of TBP-associated factors (TAFs). Interacts with TAF10 via the histone fold. Interacts with TAF13, TBP, SAP130 and GCN5L2. Interacts with TBPL2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAF3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JE64-97
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene TAF3
Gene Accession No. Q5VWG9
Gene Alias 140 kDa TATA box-binding protein-associated factor; 140kDa; 4933439M23Rik; AW539625; fi76b06; MGC25063; mTAFII140; RNA polymerase II transcription factor TAFII140; RNA polymerase II transcription factor, TFIID subunit; TAF(II)140; TAF140; Taf3; TAF3 RNA polymerase II, TATA box binding protein (TBP)-associated facto; TAF3 RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF3 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 140kDa; TAFII140; TAFII-140; TATA box binding protein associated factor 3; TATA-box binding protein associated factor 3; TBP-associated factor 3; Transcription initiation factor TFIID 140 kDa subunit; transcription initiation factor TFIID subunit 3; Unknown (protein for IMAGE:7281647); wu:fi76b06; zgc:136280
Gene Symbols TAF3
Host Species Rabbit
Immunogen Recombinant protein within human TAF3 aa 201-400/929.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83860
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.