Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAF4B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556014
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556014 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556014 Supplier Invitrogen™ Supplier No. PA556014
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PHLVPFLKKS VVALRQLLPN SQSFIQQCVQ QTSSDMVIAT CTTTVTTSPV VTTTVSSSQS EKSIIVSGAT APRTVSVQTL NPLAGPVGAK AGVVTLHSVG PTAATGGTTA GTGLLQTSKP LVTSVANTVT TVSLQPEKPV VSGTAVT Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 70%.

TAF II p105, also called TAF4B, is a celltype specific transcriptional co-activator that is a component of the TFIID complex. Expressed primarily in B cells and ovarian granulosa cells, TAF II p105 can interact with OCBA/POU2AF1 to activate B cell-specific octamerdependent transcription. Additionally, TAF II p105 plays an important role in co-activating the transcription factor NFkB and is essential for activation of anti-apoptotic genes such as TNFAIP3. TAF II p135, also known as TAF4, TAF2C, TAF2C1, TAF4A or TAFII130, is a 1,085 amino acid subunit of TFIID that accelerates transcriptional activation triggered by thyroid hormone (TR) or retinoic acid (RA). Localized to the nucleus, TAF II p135 contains one TAFH domain and is thought to bind to proteins that contain glutamine-rich domains, such as the transcription factor CREB.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAF4B
Applications ChIP Assay, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TAF4B
Gene Accession No. Q92750
Gene Alias SPGF13; TAF(II)105; TAF2C2; TAF4B; TAF4b RNA polymerase II, TATA box binding protein (TBP)-associated factor, 105kDa; TAFII105; TAFII-105; TATA box binding protein (TBP)-associated factor 4B; TATA box binding protein (TBP)-associated factor, RNA polymerase II, C2, 105kD; TATA box binding protein associated factor 4b; TATA-box binding protein associated factor 4b; transcription initiation factor TFIID 105 kDa subunit; Transcription initiation factor TFIID subunit 4B
Gene Symbols TAF4B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TAF4B. Recombinant protein control fragment (Product #RP-89354).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6875
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.