Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TAF5L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566966
Description
Immunogen sequence: NFKLRAFLDNK YVVRLQEDSY NYLIRYLQSD NNTALCKVLT LHIHLDVQPA KRTDYQLYAS GSSSRSENNG LEPPDMPSPI LQNEAALEVL QESIKRVKDG PP Highest antigen sequence identity to the following orthologs - mouse 91%, rat 91%.
TAF5L (TAF5-like RNA polymerase II p300/CBP-associated factor-associated factor 65 kDa subunit 5L), also known as PAF65B, is a 589 amino acid protein that localizes to the nucleus and contains six WD repeats. Existing as a component of the PCAF histone acetylase complex, TAF5L interacts with a number of TAFs and, via this interaction, plays a role in histone acetylation and transcription initiation. Defects in the gene encoding TAF5L, which is expressed as multiple alternatively spliced isoforms, are associated with the pathogenesis of type I diabetes.
Specifications
TAF5L | |
Polyclonal | |
Unconjugated | |
Taf5l | |
1110005N04Rik; AI849020; Paf65b; PAF65-beta; PCAF associated factor 65 beta; PCAF-associated factor 65 beta; TAF5L; TAF5-like RNA polymerase II p300/CBP-associated factor-associated factor 65 kDa subunit 5L; TAF5-like RNA polymerase II, p300/CBP-associated factor (PCAF)-associated factor; TAF5-like RNA polymerase II, p300/CBP-associated factor (PCAF)-associated factor, 65kDa; TATA box binding protein associated factor 5 like; TATA-box binding protein associated factor 5 like | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27097 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O75529 | |
Taf5l | |
Recombinant Human TAF5L. Recombinant protein control fragment (Product #RP-105441). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction