Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TAF6 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10060639
Description
TAF6 Polyclonal specifically detects TAF6 in Human samples. It is validated for Immunoprecipitation.Specifications
TAF6 | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
DKFZp781E21155, MGC:8964, RNA polymerase II, E, 70/85kD, TAF(II)70, TAF(II)80, TAF2ERNA polymerase II TBP-associated factor subunit E, TAF6 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 80kDa, TAFII70TAFII-80, TAFII80TAFII-70, TAFII85, Transcription initiation factor TFIID 70 kDa subunit, Transcription initiation factor TFIID 80 kDa subunit, transcription initiation factor TFIID subunit 6 | |
Rabbit | |
Affinity Purified | |
RUO | |
6878 | |
Store at 4C. Do not freeze. |
Immunoprecipitation | |
1.0 mg/mL | |
Immunoprecipitation 2 - 5 ug/mg lysate | |
NP_005632.1 | |
TAF6 | |
The immunogen recognized by this antibody maps to a region between residue 575 and 625 of human TBP-associated factor 6 using the numbering given in entry NP_005632.1 (GeneID 6878). | |
100 μL | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction