Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAF6 Monoclonal Antibody (25TA-2G7)
GREENER_CHOICE

Catalog No. PIMA3077
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA3077 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA3077 Supplier Invitrogen™ Supplier No. MA3077
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA3-077 detects TATA Binding protein (TBP) Associated factor 6 in human and mouse samples. MA3-077 has been successfully used in a Western blot application. The MA3-077 immunogen is a synthetic peptide coupled to ovalalbumin, from aa 39 to aa 56.

Initiation of transcription by RNA polymerase II requires the activities of more than 70 polypeptides. The protein that coordinates these activities is transcription factor IID (TFIID), which binds to the core promoter to position the polymerase properly, serves as the scaffold for assembly of the remainder of the transcription complex, and acts as a channel for regulatory signals. TFIID is composed of the TATA-binding protein (TBP) and a group of evolutionarily conserved proteins known as TBP-associated factors or TAFs. TAFs may participate in basal transcription, serve as coactivators, function in promoter recognition or modify general transcription factors (GTFs) to facilitate complex assembly and transcription initiation. This gene encodes one of the smaller subunits of TFIID that binds weakly to TBP but strongly to TAF1, the largest subunit of TFIID. Four isoforms have been identified but complete transcripts have been determined for only three isoforms. One of the isoforms has been shown to preclude binding of one of the other TFIID subunits, thereby reducing transcription and initiating signals that trigger apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAF6
Applications Western Blot
Classification Monoclonal
Clone 25TA-2G7
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.05% sodium azide
Gene TAF6
Gene Accession No. P49848, Q62311
Gene Alias 80kDa; AW549759; DKFZp781E21155; MGC:8964; p80; RNA polymerase II TBP-associated factor subunit E; RNA polymerase II, TATA box binding protein (TBP)-associated factor, 80kDa; TAF(II)70; TAF(II)80; TAF2E; Taf6; TAF6 RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF6 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 80 kDa; TAF6 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 80kDa; TAFII70; TAFII-70; TAFII80; TAFII-80; TAFII85; TATA box binding protein (Tbp)-associated factor, RNA polymerase II, E; TATA box binding protein (TBP)-associated factor, RNA polymerase II, E, 70/85kD; TATA box binding protein associated factor 6; TATA-box binding protein associated factor 6; TBP-associated factor 6; TBP-associated factor 6 isoform alpha; TFIID subunit p80; transcription initiation factor TFIID 70 kDa subunit; Transcription initiation factor TFIID 80 kDa subunit; transcription initiation factor TFIID subunit 6; wu:fb83e08; wu:fj30b02; zgc:92160
Gene Symbols TAF6
Host Species Mouse
Immunogen Synthetic peptide coupled to ovalalbumin, from aa 39 to aa 56.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21343, 6878
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.