Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAF6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541502
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541502 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541502 Supplier Invitrogen™ Supplier No. PA541502
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: PSVQPIVKLV STATTAPPST APSGPGSVQK YIVVSLPPTG EGKGGPTSHP Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 93%.

Initiation of transcription by RNA polymerase II requires the activities of more than 70 polypeptides. The protein that coordinates these activities is transcription factor IID (TFIID), which binds to the core promoter to position the polymerase properly, serves as the scaffold for assembly of the remainder of the transcription complex, and acts as a channel for regulatory signals. TFIID is composed of the TATA-binding protein (TBP) and a group of evolutionarily conserved proteins known as TBP-associated factors or TAFs. TAFs may participate in basal transcription, serve as coactivators, function in promoter recognition or modify general transcription factors (GTFs) to facilitate complex assembly and transcription initiation. This gene encodes one of the smaller subunits of TFIID that binds weakly to TBP but strongly to TAF1, the largest subunit of TFIID. Four isoforms have been identified but complete transcripts have been determined for only three isoforms. One of the isoforms has been shown to preclude binding of one of the other TFIID subunits, thereby reducing transcription and initiating signals that trigger apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAF6
Applications ChIP Assay, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TAF6
Gene Accession No. P49848
Gene Alias 80kDa; AW549759; DKFZp781E21155; MGC:8964; p80; RNA polymerase II TBP-associated factor subunit E; RNA polymerase II, TATA box binding protein (TBP)-associated factor, 80kDa; TAF(II)70; TAF(II)80; TAF2E; Taf6; TAF6 RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF6 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 80 kDa; TAF6 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 80kDa; TAFII70; TAFII-70; TAFII80; TAFII-80; TAFII85; TATA box binding protein (Tbp)-associated factor, RNA polymerase II, E; TATA box binding protein (TBP)-associated factor, RNA polymerase II, E, 70/85kD; TATA box binding protein associated factor 6; TATA-box binding protein associated factor 6; TBP-associated factor 6; TBP-associated factor 6 isoform alpha; TFIID subunit p80; transcription initiation factor TFIID 70 kDa subunit; Transcription initiation factor TFIID 80 kDa subunit; transcription initiation factor TFIID subunit 6; wu:fb83e08; wu:fj30b02; zgc:92160
Gene Symbols TAF6
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human TAF6 (aa 576-625).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6878
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.