Learn More
Invitrogen™ TAF7 Polyclonal Antibody

Description
Recommended positive controls: A431, JurkatHeLa, HepG2, A431, Jurkat. Predicted reactivity: Mouse (89%), Rat (91%), Chicken (86%), Rhesus Monkey (100%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | TAF7 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.8 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | TAF7 |
| Gene Accession No. | Q15545 |
| Gene Alias | 55kDa; AI607308; BB007175; RNA polymerase II TBP-associated factor subunit F; TAF(II)55; TAF2F; Taf7; TAF7 RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF7 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 55 kDa; TAF7 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 55kDa; TAFII55; TAFII-55; TATA box binding protein (TBP) associated factor, RNA polymerase II, F, 55 kDa; TATA box binding protein (TBP)-associated factor, RNA polymerase II, F, 55kD; TATA box binding protein associated factor 7; TATA-box binding protein associated factor 7; TBP-associated factor F; transcription factor IID subunit TAFII55; transcription initiation factor TFIID 55 kDa subunit; Transcription initiation factor TFIID subunit 7; transcription initiation factor TFIID, 55 kDa subunit |
| Gene Symbols | TAF7 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.