Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAF7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552296
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552296 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552296 Supplier Invitrogen™ Supplier No. PA552296
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ISSPGMSGHR QGHDSLEHDE LREIFNDLSS SSEDEDETQH QDEEDINIID TEEDLERQLQ DKLNESDEQH QENEGTNQLV MGIQKQIDNM KGKLQETQDR AKRQEDLIMK VENLALKN Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.

The intronless gene for this transcription coactivator is located between the protocadherin beta and gamma gene clusters on chromosome 5. The protein encoded by this gene is a component of the TFIID protein complex, a complex which binds to the TATA box in class II promoters and recruits RNA polymerase II and other factors. This particular subunit interacts with the largest TFIID subunit, as well as multiple transcription activators. The protein is required for transcription by promoters targeted by RNA polymerase II. The general transcription factor, TFIID, consists of the TATA-binding protein (TBP) associated with a series of TBP-associated factors (TAFs) that together participate in the assembly of the transcription preinitiation complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAF7
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TAF7
Gene Accession No. Q15545
Gene Alias 55kDa; AI607308; BB007175; RNA polymerase II TBP-associated factor subunit F; TAF(II)55; TAF2F; Taf7; TAF7 RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF7 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 55 kDa; TAF7 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 55kDa; TAFII55; TAFII-55; TATA box binding protein (TBP) associated factor, RNA polymerase II, F, 55 kDa; TATA box binding protein (TBP)-associated factor, RNA polymerase II, F, 55kD; TATA box binding protein associated factor 7; TATA-box binding protein associated factor 7; TBP-associated factor F; transcription factor IID subunit TAFII55; transcription initiation factor TFIID 55 kDa subunit; Transcription initiation factor TFIID subunit 7; transcription initiation factor TFIID, 55 kDa subunit
Gene Symbols TAF7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TAF7. Recombinant protein control fragment (Product #RP-89412).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6879
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.