Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAF7L Monoclonal Antibody (3E10)
GREENER_CHOICE

Catalog No. PIMA524476
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524476 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524476 Supplier Invitrogen™ Supplier No. MA524476
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: KKRFRKTQKK VPDVKEMEKS SFTEYIESPD VENEVKRLLR SDAEAVSTRW EVIAEDGTKE IESQGSIPGF LISSGMSSHK QGHTSSEYDM LREMFSDSRS NN.

Probably functions as a spermatogensis-specific component of the DNA-binding general transcription factor complex TFIID, a multimeric protein complex that plays a central role in mediating promoter responses to various activators and repressors. May play a role in spermatogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAF7L
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 3E10
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene TAF7L
Gene Accession No. Q5H9L4
Gene Alias 4933438I11Rik; 50kDa; cancer/testis antigen 40; CT40; RGD1564898; RNA polymerase II TBP-associated factor subunit Q; TAF2Q; Taf7l; TAF7-like RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF7-like RNA polymerase II, TATA box binding protein (TBP)-associated factor, 50kDa; TATA box binding protein (TBP)-associated factor, RNA polymerase II, Q; TATA box binding protein associated factor 7 like; TATA box binding protein-associated factor RNA polymerase II subunit Q; TATA box-binding protein-associated factor 50 kDa; TATA-box binding protein associated factor 7 like; TBP-associated factor, RNA polymerase II, Q; transcription initiation factor TFIID 50 kDa subunit; Transcription initiation factor TFIID subunit 7-like
Gene Symbols TAF7L
Host Species Mouse
Immunogen TAF7L (NP_079161.2, 231 a.a. ~ 332 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54457
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.