Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAF7L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142502
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142502 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142502 Supplier Thermo Scientific Supplier No. PA5142502
Add to Cart
Add to Cart
This item is not returnable. View return policy

TAF7L Polyclonal Antibody

TAF7L Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen TAF7L
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene TAF7L
Gene Accession No. Q5H9L4
Gene Alias 4933438I11Rik; 50kDa; cancer/testis antigen 40; CT40; RGD1564898; RNA polymerase II TBP-associated factor subunit Q; TAF2Q; Taf7l; TAF7-like RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF7-like RNA polymerase II, TATA box binding protein (TBP)-associated factor, 50kDa; TATA box binding protein (TBP)-associated factor, RNA polymerase II, Q; TATA box binding protein associated factor 7 like; TATA box binding protein-associated factor RNA polymerase II subunit Q; TATA box-binding protein-associated factor 50 kDa; TATA-box binding protein associated factor 7 like; TBP-associated factor, RNA polymerase II, Q; transcription initiation factor TFIID 50 kDa subunit; Transcription initiation factor TFIID subunit 7-like
Gene Symbols TAF7L
Host Species Goat
Immunogen Peptide with sequence C-NHFQSVLEQLELQEK (aa 429-443).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54457
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.