Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TAF8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA548572
Description
Predicted to react with bovine and chicken based on sequence homology.
Transcription factor TFIID is one of the general factors required for accurate and regulated initiation by RNA polymerase II. Mediates both basal and activator-dependent transcription. Plays a role in the differentiation of preadipocyte fibroblasts to adipocytes, however, does not seem to play a role in differentiation of myoblasts. Required for the integration of TAF10 in the TAF complex. May be important for survival of cells of the inner cell mass which constitute the pluripotent cell population of the early embryo.
Specifications
TAF8 | |
Polyclonal | |
Unconjugated | |
TAF8 | |
AW260255; hTAFII43; II; Protein taube nuss; Protein taube nuss-like; similar to taube nuss; TAF; TAF(II)43; TAF8; TAF8 RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF8 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 43kDa; TAF8 RNA polymerase II, TATA box binding protein (TBP)-associated factorq; TAFII43; TAFII-43; TATA box binding protein (TBP)-associated factor, RNA polymerase II, A, 45/50kDa; TATA box binding protein associated factor 8; TATA-box binding protein associated factor 8; taube nuss; taube nuss homolog; TBN; tbnl; tbnl protein; TBP-associated factor 43 kDa; TBP-associated factor 8; TBP-associated factor TAFII43; TBP-associated factor, RNA polymerase II, 43 kD; transcription initiation factor TFIID 43 kDa subunit; Transcription initiation factor TFIID subunit 8; zgc:56401; zgc:77837 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
129685 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q7Z7C8 | |
TAF8 | |
KLH conjugated synthetic peptide between 151-178 amino acids from the Central region of human TAF8. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction