Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAF8 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501564486
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-4486 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-4486 Supplier Bethyl Laboratories, Inc Supplier No. A303186AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse and Bovine

Transcription factor TFIID is one of the general factors required for accurate and regulated initiation by RNA polymerase II. Mediates both basal and activator-dependent transcription. Plays a role in the differentiation of preadipocyte fibroblasts to adipocytes, however, does not seem to play a role in differentiation of myoblasts. Required for the integration of TAF10 in the TAF complex. May be important for survival of cells of the inner cell mass which constitute the pluripotent cell population of the early embryo.

Specifications

Antigen TAF8
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene TAF8
Gene Accession No. Q7Z7C8
Gene Alias Transcription initiation factor TFIID subunit 8, Protein taube nuss, TBP-associated factor 43 kDa, TBP-associated factor 8, Transcription initiation factor TFIID 43 kDa subunit, TAFII-43, TAFII43, hTAFII43, TAF8, TAFII43, TBN
Gene Symbols TAF8
Host Species Rabbit
Immunogen Between 260 and 310
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 129685
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.