Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TAF9 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$196.10 - $499.50
Specifications
Antigen | TAF9 |
---|---|
Concentration | 1 mg/mL |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
TAF9 Polyclonal specifically detects TAF9 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
TAF9 | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
AD-004, adenylate kinase 6, adenylate kinase isoenzyme 6, adrenal gland protein AD-004, AK6CIP, ATP-AMP transphosphorylase 6, CGI-137, CINAPMGC:1603, coilin interacting protein, Coilin-interacting nuclear ATPase protein, EC 2.7.4.3, hCINAP, MGC1603, MGC3647, MGC5067, RNA polymerase II TBP-associated factor subunit G, STAF31/32, TAF2GMGC:5067, TAF9 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 32kDa, TAFII-31, TAFII31MGC:3647, TAFII-32, TAFII32TATA box binding protein (TBP)-associated factor, RNA polymerase II, G, 32kD, TAFIID32, transcription initiation factor TFIID 31 kD subunit, Transcription initiation factor TFIID 31 kDa subunit, Transcription initiation factor TFIID 32 kDa subunit, transcription initiation factor TFIID subunit 9 | |
TAF9 | |
IgG | |
Affinity Purified | |
28 kDa |
1 mg/mL | |
Polyclonal | |
Rabbit | |
DNA replication Transcription Translation and Splicing | |
PBS with 0.02% Sodium Azide | |
6880 | |
Antibody was raised against a 17 amino acid peptide near the center of human TAF9. The immunogen is located within amino acids 110 - 160 of TAF9. Amino Acid Squence: LTAPNYRLKSLQKKAS | |
Primary | |
Multiple isoforms of TAF9 are known to exist. TAF9 is predicted to not cross-react with other TAF family members. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title