Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAF9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540919
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540919 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540919 Supplier Invitrogen™ Supplier No. PA540919
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: GLKYINVGDL AREEQLYDGY DEEYDCPILD EDRVVDELDN QMREGGVIVD Sequence homology: Cow: 93%; Guinea Pig: 79%; Horse: 86%; Human: 100%; Mouse: 86%; Pig: 93%; Rabbit: 79%; Rat: 90%.

Initiation of transcription by RNA polymerase II requires the activities of more than 70 polypeptides. The protein that coordinates these activities is transcription factor IID (TFIID), which binds to the core promoter to position the polymerase properly, serves as the scaffold for assembly of the remainder of the transcription complex, and acts as a channel for regulatory signals. TFIID is composed of the TATA-binding protein (TBP) and a group of evolutionarily conserved proteins known as TBP-associated factors or TAFs. TAFs may participate in basal transcription, serve as coactivators, function in promoter recognition or modify general transcription factors (GTFs) to facilitate complex assembly and transcription initiation. This gene encodes one of the smaller subunits of TFIID that binds to the basal transcription factor GTF2B as well as to several transcriptional activators such as p53 and VP16. A similar but distinct gene (TAF9L) has been found on the X chromosome and a pseudogene has been identified on chromosome 19. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAF9
Applications ChIP Assay, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TAF9
Gene Accession No. Q16594
Gene Alias 2310012M09Rik; AA673500; AD-004; AU045423; CGI-137; CINAP; CIP; EMBL:AAH91109.1}; hCINAP; MGC:1603; MGC:3647; MGC:5067; RNA polymerase II TBP-associated factor subunit G; STAF31/32; TAF2G; TAF9; taf9 {ECO:0000312; TAF9 RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF9 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 32 kDa; TAF9 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 32kDa; TAF9 RNA polymerase II, TATA box binding protein-associated factor; Tafii31; TAFII-31; TAFII32; TAFII-32; TAFIID32; TATA box binding protein associated factor 9; TATA-box binding protein associated factor 9; transcription initiation factor TFIID 31 kD subunit; transcription initiation factor TFIID 31 kDa subunit; Transcription initiation factor TFIID 32 kDa subunit; transcription initiation factor TFIID subunit 9
Gene Symbols TAF9
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human TAF9.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6880
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.