Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAF9B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103026
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103026 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103026 Supplier Invitrogen™ Supplier No. PA5103026
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TAF9B.

The protein encoded by this gene is a methylcytosine dioxygenase that catalyzes the conversion of methylcytosine to 5-hydroxymethylcytosine. The encoded protein is involved in myelopoiesis, and defects in this gene have been associated with several myeloproliferative disorders. Two variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAF9B
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Taf9b
Gene Accession No. Q6NZA9, Q9HBM6
Gene Alias BC066223; Dn7; DN-7; neuronal cell death-related gene in neuron 7; neuronal cell death-related gene in neuron-7; neuronal cell death-related protein 7; Taf9b; TAF9B RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF9B RNA polymerase II, TATA box binding protein (TBP)-associated factor, 31kDa; TAF9L; TAF9-like RNA polymerase II TATA box binding protein (TBP)-associated factor 31 kD; TAF9-like RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF9-like RNA polymerase II, TATA box binding protein (TBP)-associated factor, 31 kD; TAF9-like RNA polymerase II, TATA box binding protein (TBP)-associated factor, 31kDa; TAFII31; TAFII31L; TATA box binding protein associated factor 9b; TATA-box binding protein associated factor 9b; TBP-associated factor 9L; TFIID-31; transcription associated factor TAFII31L; transcription initiation factor IID, 31kD subunit; Transcription initiation factor TFIID subunit 9B; Transcription initiation factor TFIID subunit 9-like; transcription-associated factor TAFII31L
Gene Symbols Taf9b
Host Species Rabbit
Immunogen A synthesized peptide derived from human TAF9B(Accession Q9HBM6), corresponding to amino acid residues M1-Q38.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 407786, 51616
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.