Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TAF9B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571344
Description
TAF9B Polyclonal Antibody for Western Blot
The protein encoded by this gene is a methylcytosine dioxygenase that catalyzes the conversion of methylcytosine to 5-hydroxymethylcytosine. The encoded protein is involved in myelopoiesis, and defects in this gene have been associated with several myeloproliferative disorders. Two variants encoding different isoforms have been found for this gene.
Specifications
TAF9B | |
Polyclonal | |
Unconjugated | |
Taf9b | |
BC066223; Dn7; DN-7; neuronal cell death-related gene in neuron 7; neuronal cell death-related gene in neuron-7; neuronal cell death-related protein 7; Taf9b; TAF9B RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF9B RNA polymerase II, TATA box binding protein (TBP)-associated factor, 31kDa; TAF9L; TAF9-like RNA polymerase II TATA box binding protein (TBP)-associated factor 31 kD; TAF9-like RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF9-like RNA polymerase II, TATA box binding protein (TBP)-associated factor, 31 kD; TAF9-like RNA polymerase II, TATA box binding protein (TBP)-associated factor, 31kDa; TAFII31; TAFII31L; TATA box binding protein associated factor 9b; TATA-box binding protein associated factor 9b; TBP-associated factor 9L; TFIID-31; transcription associated factor TAFII31L; transcription initiation factor IID, 31kD subunit; Transcription initiation factor TFIID subunit 9B; Transcription initiation factor TFIID subunit 9-like; transcription-associated factor TAFII31L | |
Rabbit | |
Affinity chromatography | |
RUO | |
51616 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9HBM6 | |
Taf9b | |
synthetic peptide directed towards the C-terminal region of human TAF9B. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction