Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAFA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571440
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571440 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571440 Supplier Invitrogen™ Supplier No. PA571440
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TAFA2 Polyclonal Antibody for Western Blot

This gene is a member of the TAFA family which is composed of five highly homologous genes that encode small secreted proteins. These proteins contain conserved cysteine residues at fixed positions, and are distantly related to MIP-1alpha, a member of the CC-chemokine family. The TAFA proteins are predominantly expressed in specific regions of the brain, and are postulated to function as brain-specific chemokines or neurokines, that act as regulators of immune and nervous cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAFA2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FAM19A2
Gene Accession No. Q8N3H0
Gene Alias Chemokine-like protein TAFA-2; FAM19A2; family with sequence similarity 19 (chemokine (C-C motif)-like), member A2; family with sequence similarity 19 member A2, C-C motif chemokine like; protein FAM19A2; TAFA2; TAFA-2
Gene Symbols FAM19A2
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human TAFA2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 338811
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.