Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TAFA3/FAM19A3 Antibody (460904), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB4677B
Description
TAFA3/FAM19A3 Monoclonal antibody specifically detects TAFA3/FAM19A3 in Human samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), CyTOFSpecifications
TAFA3/FAM19A3 | |
Monoclonal | |
Biotin | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
284467 | |
Store at 4°C in the dark. | |
IgG2b |
Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
460904 | |
Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
Chemokine-like protein TAFA-3, family with sequence similarity 19 (chemokine (C-C motif)-like), member A3, MGC138473, TAFA3, TAFA-3 | |
E. coli-derived recombinant human TAFA3/FAM19A3, Ala31-Arg133, Accession # Q7Z5A8 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction