Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAFA3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563210
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563210 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563210 Supplier Invitrogen™ Supplier No. PA563210
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AALQPPTATV LVQQGTCEVI A Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 86%.

The TAFA3 gene encodes a protein that is part of the TAFA family of chemokine-like proteins, involved in modulating immune responses and inflammatory processes. TAFA3 is particularly implicated in the regulation of neuroinflammatory and neuroprotective activities in the central nervous system. It functions as a signaling molecule that can influence cell survival, proliferation, and migration, contributing to homeostasis in neural tissue and promoting recovery following injury. TAFA3 has been studied for its role in neuroprotection, where it helps shield neurons from damage caused by oxidative stress and inflammatory stimuli, making it significant in neurodegenerative conditions. Apart from its neuroprotective capabilities, TAFA3 may also be involved in the systemic modulation of inflammation, affecting immune cell trafficking and responses. Research into TAFA3 continues to unveil its multifaceted role in health and disease, highlighting potential therapeutic avenues for targeting this protein in conditions characterized by excessive inflammation or neurodegeneration.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAFA3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM19A3
Gene Accession No. Q7Z5A8
Gene Alias Chemokine-like protein TAFA-3; FAM19A3; family with sequence similarity 19 (chemokine (C-C motif)-like), member A3; family with sequence similarity 19 member A3, C-C motif chemokine like; protein FAM19A3; TAFA3; TAFA-3
Gene Symbols FAM19A3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TAFA3. Recombinant protein control fragment (Product #RP-101557).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284467
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.