Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TAFA4/FAM19A4 Antibody (480103), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB5099A
Description
TAFA4/FAM19A4 Monoclonal antibody specifically detects TAFA4/FAM19A4 in Human samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), CyTOFSpecifications
TAFA4/FAM19A4 | |
Monoclonal | |
APC | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
151647 | |
Store at 4°C in the dark. | |
IgG1 |
Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
480103 | |
Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
Chemokine-like protein TAFA-4, family with sequence similarity 19 (chemokine (C-C motif)-like), member A4, FLJ25161, TAFA4, TAFA-4 | |
E. coli-derived recombinant human TAFA4, Ser35-Arg140, Accession # Q96LR4 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction