Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TAFA4/FAM19A4 Antibody (480103), CoraFluor™ 1, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB5099CL1
Description
TAFA4/FAM19A4 Monoclonal antibody specifically detects TAFA4/FAM19A4 in Human samples. It is validated for Western Blot,Flow Cytometry (Intracellular Staining),CyTOFSpecifications
| TAFA4/FAM19A4 | |
| Monoclonal | |
| CoraFluor 1 | |
| Chemokine-like protein TAFA-4, family with sequence similarity 19 (chemokine (C-C motif)-like), member A4, FLJ25161, TAFA4, TAFA-4 | |
| E. coli-derived recombinant human TAFA4, Ser35-Arg140, Accession # Q96LR4 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
| 480103 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 151647 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction