Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TAFA4/FAM19A4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP159651
Description
TAFA4/FAM19A4 Polyclonal specifically detects TAFA4/FAM19A4 in Human samples. It is validated for Western Blot.Specifications
| TAFA4/FAM19A4 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| Chemokine-like protein TAFA-4, family with sequence similarity 19 (chemokine (C-C motif)-like), member A4, FLJ25161, TAFA4, TAFA-4 | |
| Rabbit | |
| 36 kDa | |
| 100 μL | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q96LR4 | |
| FAM19A4 | |
| Synthetic peptides corresponding to FAM19A4 (family with sequence similarity 19 (chemokine (C-C motif)-like), member A4) The peptide sequence was selected from the middle region of FAM19A4)(50ug). Peptide sequence SSQHLRGHAGHHQIKQGTCEVVAVHRCCNKNRIEERSQTVKCSCFPGQVA The peptide sequence for this immunogen was taken from within the described region. | |
| Affinity purified | |
| RUO | |
| 151647 | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction