Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAFA5 Monoclonal Antibody (463102), Invitrogen™

Rat Monoclonal Antibody

Supplier:  Thermo Scientific MA524183

Catalog No. PIMA524183


Only null left
Add to Cart

Description

Description

In direct ELISAs and Western blots, no cross-reactivity with recombinant human TAFA2, 3, or 4 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

TAFA5 Monoclonal antibody specifically detects TAFA5 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TAFA5
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
Q7Z5A7
TAFA5
E. coli-derived recombinant human TAFA5/FAM19A5 Gln26-Ser125
100 μg
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
463102
Unconjugated
TAFA5
Chemokine-like protein TAFA-5; FAM19A5; family with sequence similarity 19 (chemokine (C-C motif)-like), member A5; family with sequence similarity 19 member A5, C-C motif chemokine like; family with sequence similarity 19, member A5; protein FAM19A5; QLLK5208; RGD1562115; TAFA protein 5; TAFA5; Tafa-5; Tara-5; UNQ5208; UNQ5208/PRO34524
Rat
Protein A/G
RUO
25817
-20° C, Avoid Freeze/Thaw Cycles
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.