Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TAFA5 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Supplier: Thermo Scientific PA547702
Description
In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) TAFA1, rhTAFA2, rhTAFA3, and rhTAFA4 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
TAFA5 Polyclonal antibody specifically detects TAFA5 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
TAFA5 | |
Polyclonal | |
Unconjugated | |
TAFA5 | |
Chemokine-like protein TAFA-5; FAM19A5; family with sequence similarity 19 (chemokine (C-C motif)-like), member A5; family with sequence similarity 19 member A5, C-C motif chemokine like; family with sequence similarity 19, member A5; protein FAM19A5; QLLK5208; RGD1562115; TAFA protein 5; TAFA5; Tafa-5; Tara-5; UNQ5208; UNQ5208/PRO34524 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
106014, 25817, 500915 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q7Z5A7, Q91WE9 | |
TAFA5 | |
E. coli-derived recombinant human TAFA5/FAM19A5 Gln26-Ser125 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction