Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAFII p135/p105 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA536243
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA536243 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA536243 Supplier Invitrogen™ Supplier No. PA536243
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 135 kDa and 105 kDa. Purity is >95% by SDS-PAGE.

TAF II p105, also called TAF4B, is a celltype specific transcriptional co-activator that is a component of the TFIID complex. Expressed primarily in B cells and ovarian granulosa cells, TAF II p105 can interact with OCBA/POU2AF1 to activate B cell-specific octamerdependent transcription. Additionally, TAF II p105 plays an important role in co-activating the transcription factor NFkB and is essential for activation of anti-apoptotic genes such as TNFAIP3. TAF II p135, also known as TAF4, TAF2C, TAF2C1, TAF4A or TAFII130, is a 1,085 amino acid subunit of TFIID that accelerates transcriptional activation triggered by thyroid hormone (TR) or retinoic acid (RA). Localized to the nucleus, TAF II p135 contains one TAFH domain and is thought to bind to proteins that contain glutamine-rich domains, such as the transcription factor CREB.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAFII p135/p105
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene TAF4
Gene Accession No. G5E8Z2, O00268, Q92750
Gene Alias 105kDa; 135kDa; 2610524B04Rik; 4932409F03Rik; AI450312; AW987595; RGD1562997; RNA polymerase II TBP-associated factor subunit C; similar to Transcription initiation factor TFIID 105 kDa subunit (TAFII-105); SPGF13; TAF(II)105; TAF(II)130; TAF(II)135; TAF2C; TAF2C1; Taf2c2; TAF4; TAF4 RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF4 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 135kDa; Taf4a; TAF4A RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF4A RNA polymerase II, TATA box binding protein (TBP)-associated factor, 135 kD; Taf4b; TAF4B RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF4B RNA polymerase II, TATA box binding protein (TBP)-associated factor, 105 kDa; TAF4b RNA polymerase II, TATA box binding protein (TBP)-associated factor, 105kDa; TAFII 130; TAFII 135; TAFII105; TAFII-105; TAFII130; TAFII-130; TAFII135; TAFII-135; TATA box binding protein (TBP)-associated factor 4B; TATA box binding protein (TBP)-associated factor, RNA polymerase II, C1, 130kd; TATA box binding protein (TBP)-associated factor, RNA polymerase II, C2, 105kD; TATA box binding protein associated factor 4; TATA box binding protein associated factor 4b; TATA-box binding protein associated factor 4; TATA-box binding protein associated factor 4b; TBP-associated factor 4; Transcription initiation factor TFIID 105 kDa subunit; transcription initiation factor TFIID 130 kDa subunit; transcription initiation factor TFIID 135 kD subunit; transcription initiation factor TFIID 135 kDa subunit; Transcription initiation factor TFIID subunit 4; transcription initiation factor TFIID subunit 4B
Gene Symbols TAF4, TAF4B
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 951-1000 of human TAF II p135.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 228980, 291773, 682097, 6874, 6875, 72504
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.