Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TAG-72 Antibody (B72.3 (Satumomab)) - Chimeric - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP2526720.2MG
Description
TAG-72 Monoclonal specifically detects TAG-72 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, CyTOF-ready.Specifications
TAG-72 | |
Monoclonal | |
Unconjugated | |
PBS with 0.02% Proclin 300. with 0.02% Proclin 300 | |
Rabbit | |
Protein A or G purified | |
RUO | |
182875 | |
Human | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, CyTOF | |
B72.3 (Satumomab) | |
Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, CyTOF-ready | |
CA 72-4, Protein TAG-72, tag 72, tag-72 Protein TAG-72 | |
Tumour-associated glycoprotein 72 is a glycoprotein found on the surface of many cancer cells, including ovary, breast, colon, and pancreatic cells. | |
0.2 mg | |
Primary | |
Recognises human Tumor associated glycoprotein 72. | |
Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction