Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TAG-72 Rabbit anti-Human, Rat, Bovine, Canine, Hamster, Clone: CA72/2869R, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP30769920UG
Description
TAG-72 Monoclonal specifically detects TAG-72 in Human, Rat, Bovine, Canine, Hamster samples. It is validated for Immunohistochemistry-Paraffin.Specifications
TAG-72 | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 ug/ml | |
CA 72-4, Protein TAG-72, tag 72, tag-72 Protein TAG-72 | |
Purified TAG-72 protein | |
20 μg | |
Primary | |
Store at 4°C. | |
IgG |
Immunohistochemistry (Paraffin) | |
CA72/2869R | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Rabbit | |
Protein A or G purified | |
RUO | |
Human, Rat, Bovine, Canine, Hamster | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction