Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAG72 Monoclonal Antibody (EBS-T-059)
GREENER_CHOICE

Catalog No. PIMA533331
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA533331 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA533331 Supplier Invitrogen™ Supplier No. MA533331
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EBS-T-059 recognizes an oncofetal antigen of 220kDa, identified as a tumor-associated glycoprotein (TAG72) with properties of a mucin and usually expressed by adenocarcinomas, but not by mesotheliomas and thus helps in the differential diagnosis of malignancies of the lung. The combined use of anti-TAG-72 and anti-GCDFP-15 is valuable in the diagnosis of apocrine carcinoma. TAG72 is further used as serum marker for gastric cancer.

Tumor-associated glycoprotein (TAG-72) is an oncofetal antigen with properties of a mucin. About 60% of carcinoma patients express TAG-72 in their sera.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAG72
Applications ELISA, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone EBS-T-059
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Alias TAG-72; Tumor-associated glycoprotein 72
Host Species Mouse
Immunogen Human TAG72 recombinant protein.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.