Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Tagged Column Package, His (2-step Purification Protocol)

Catalog No. 09929012
Click to view available options
Quantity:
1 Kit

Combines affinity with size exclusion chromatography to obtain highly pure his-tagged proteins

  • Contains a HisTrap™ HP column (1 mL) prepacked with Ni Sepharose High Performance (HP) affinity resin for preparative purification of his-tagged proteins
  • Achieve final purity with HiPrep™ 16/60 Sephacryl S-200 HR size exclusion chromatography column, which removes any remaining trace of impurities or closely related substances and imidazole
  • Includes instructions with guidance and tips on how to get started
TRUSTED_SUSTAINABILITY

Specifications

Kit Contents HisTrap™ HP Column Prepacked with Ni Sepharose High Performance (HP) Affinity Resin, and HiPrep™ 16/60 Sephacryl S-200 HR Size Exclusion Chromatography Column
Content And Storage 4°C to 30°C
Quantity 1 Kit
For Use With (Application) Purification of His-Tagged Proteins
Type Protein Purification System
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.