Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAGLN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518252
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518252 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518252 Supplier Invitrogen™ Supplier No. PA518252
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine and canine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.

TAGLN is a transformation and shape-change sensitive actin cross-linking/gelling protein found in fibroblasts and smooth muscle. Its expression is down-regulated in many cell lines, and this down-regulation may be an early and sensitive marker for the onset of transformation. A functional role of this protein is unclear.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAGLN
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene TAGLN
Gene Accession No. Q01995
Gene Alias 22 kDa actin-binding protein; 25 kDa F-actin-binding protein; actin-associated protein p27; DKFZp686B01212; DKFZp686P11128; Protein WS3-10; similar to transgelin; Sm22; sm22 alpha; SM22A; Sm22alpha; SM22-alpha; SMCC; smooth muscle 22 protein; smooth muscle protein 22-alpha; TAGLN; TAGLN1; Transgelin; transgelin variant 2; Ws310; WS3-10
Gene Symbols TAGLN
Host Species Goat
Immunogen Synthetic peptide sequence (MTGYGRPRQIIS) corresponding to the C-terminus amino acids of TAGLN (aa 190-201).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6876
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.