Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAGLN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529767
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529767 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529767 Supplier Invitrogen™ Supplier No. PA529767
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, HeLa, HepG2, Mouse liver, A-10, A-10 treated with MG132, Bovine Smooth Muscle Cell, Bovine Fibroblastic Chondrocyte. Predicted reactivity: Mouse (97%), Rat (96%), Xenopus laevis (81%), Pig (98%), Chicken (87%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

TAGLN is a transformation and shape-change sensitive actin cross-linking/gelling protein found in fibroblasts and smooth muscle. Its expression is down-regulated in many cell lines, and this down-regulation may be an early and sensitive marker for the onset of transformation. A functional role of this protein is unclear.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAGLN
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Gene TAGLN
Gene Accession No. P31232, P37804, Q01995, Q9TS87
Gene Alias 22 kDa actin-binding protein; 25 kDa F-actin-binding protein; actin-associated protein p27; DKFZp686B01212; DKFZp686P11128; Protein WS3-10; similar to transgelin; Sm22; sm22 alpha; SM22A; Sm22alpha; SM22-alpha; SMCC; smooth muscle 22 protein; smooth muscle protein 22-alpha; TAGLN; TAGLN1; Transgelin; transgelin variant 2; Ws310; WS3-10
Gene Symbols TAGLN
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Transgelin. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21345, 25123, 513463, 6876
Target Species Bovine, Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.