Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAGLN3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564039
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564039 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564039 Supplier Invitrogen™ Supplier No. PA564039
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EQKYDADLEN KLVDWIILQC AEDIEHPP Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

The transgelin family is a group of proteins that belong to 22 kDa actin-related corpnin superfamily. Of all three isoforms, transgelin 1 is the best characterized. Transgelin 1, also known as SM22alpha, is a specific marker for differentiated smooth muscle cells. Transgelin 2, also known as SM22 beta, is expressed by both smooth muscle and non-smooth muscle cells in a temporally and spatially regulated pattern. Trangenlin 3, also known as NP25, is only found in highly differentiated neuronal cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAGLN3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TAGLN3
Gene Accession No. Q9UI15
Gene Alias 2700038H05Rik; 2900005O10Rik; AI426007; neuronal protein 22; Neuronal protein NP25; NP22; NP24; NP25; Tagln3; transgelin 3; transgelin-3
Gene Symbols TAGLN3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TAGLN3. Recombinant protein control fragment (Product #RP-103458).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29114
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.