Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAIP12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518657
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518657 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518657 Supplier Invitrogen™ Supplier No. PA518657
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with canine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.

The protein encoded by this gene belongs to the CSRNP family of nuclear proteins that share conserved regions, including cysteine- and serine- rich regions, a basic domain, a transcriptional activation domain, and bind the sequence 'AGAGTG', thus have the hallmark of transcription factors. Studies in mice suggest that these genes may have redundant functions. Alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAIP12
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Csrnp2
Gene Accession No. Q8BGQ2, Q9H175
Gene Alias BC035295; C12orf2; C12orf22; Csnrp2; CSRN2; Csrnp2; CSRNP-2; cysteine and serine rich nuclear protein 2; cysteine/serine-rich nuclear protein 2; cysteine-serine-rich nuclear protein 2; Fam130a1; family with sequence similarity 130, member A1; FLJ25576; PPP1R72; Protein FAM130A1; protein phosphatase 1, regulatory subunit 72; RGD1308916; TAIP12; TAIP-12; TGF-beta induced apoptosis protein 12; TGF-beta-induced apoptosis protein 12
Gene Symbols Csrnp2
Host Species Goat
Immunogen Synthetic peptide sequence (KRQKQLRRKNVRFDQ) corresponding to the internal amino acids of BC035295 (aa 56-70).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 207785, 81566
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.