Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAIP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102010
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102010 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102010 Supplier Invitrogen™ Supplier No. PA5102010
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TAIP-2.

TAIP2 binds to the consensus sequence 5'-AGAGTG-3' and has transcriptional activator activity. Plays a role in apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAIP2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CSRNP3
Gene Accession No. P59055, Q8WYN3
Gene Alias A330102K23Rik; Csnrp3; Csrnp3; CSRNP-3; cysteine and serine rich nuclear protein 3; cysteine/serine-rich nuclear protein 3; cysteine-serine-rich nuclear protein 3; Fam130a2; family with sequence similarity 130, member A2; Mbu1; PPP1R73; Protein FAM130A2; protein phosphatase 1, regulatory subunit 73; RGD1306214; TAIP2; TAIP-2; TGF beta induced apotosis protein 2; TGF-beta induced apoptosis protein 2; TGF-beta-induced apoptosis protein 2
Gene Symbols CSRNP3
Host Species Rabbit
Immunogen A synthesized peptide derived from human CSRNP3(Accession Q8WYN3), corresponding to amino acid residues F173-D223.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 311093, 77771, 80034
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.