Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TAK 165
GREENER_CHOICESDP

Catalog No. 359910R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-991-0R 10 mg
35-995-0R 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 35-991-0R Supplier Tocris Bioscience™ Supplier No. 3599/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective ErbB2 inhibitor

TAK 165 is a potent, irreversible human epithelial growth factor receptor 2 (ErbB2) inhibitor (IC50 = 6 nM) that displays > 4000-fold selectivity over EGFR, FGFR, PDGFR, JAK1 and Src. Exhibits potent antiproliferative effects in ErbB2-overexpressing cancer cell lines (IC50 = 5 nM in BT474 breast cancer cells) and significantly inhibits bladder, breast and prostate cancer xenograft growth in vivo.

Specifications

Quantity 10 mg
Product Type TAK 165
Purity >99%
Molecular Weight (g/mol) 468.47
Inhibitors TAK 165

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.