Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TAK1, p Ser439 Antibody (SR2176), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | TAK1 |
---|---|
Clone | SR2176 |
Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 1:50 |
Applications | Western Blot, Immunoprecipitation |
Classification | Monoclonal |
Description
TAK1 Monoclonal antibody specifically detects TAK1 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
TAK1 | |
Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
EC 2.7.11, EC 2.7.11.25, MEKK7, mitogen-activated protein kinase kinase kinase 7, TAK1/MAP3K7, TAK1TGF1a, TGF-beta activated kinase 1, TGF-beta-activated kinase 1, Transforming growth factor-beta-activated kinase 1 | |
A synthesized peptide derived from human phospho-TAK1 [Ser439] (Uniprot #: O43318) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR2176 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Apoptosis, Immunology, Innate Immunity, Phospho Specific, Protein Kinase, Signal Transduction, Wnt Signaling Pathway | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
6885 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title