Learn More
Promega TAK1-TAB1 Kinase Enzyme System, 1mg
Catalog No. PRV6564 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6564 1 mg
1 options
Catalog No. PRV6564 Supplier Promega Supplier No. V6564
Add to Cart
Add to Cart
Request a Quote
Recombinant human proteins TAK1 (1-303) and TAB1 (437-end), linked by a small peptide (DFGGGGG), were expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Easily Screen and Profile TAK1−TAB1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C