Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAL1 Monoclonal Antibody (OTI5A5), TrueMAB™, OriGene
SDP

Catalog No. 502540648
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0648 30 μL
50-167-4676 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0648 Supplier OriGene Supplier No. TA503898S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TAL1 Monoclonal antibody specifically detects TAL1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot

Specifications

Antigen TAL1
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI5A5
Concentration 0.87 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene TAL1
Gene Accession No. P17542
Gene Alias basic helix-loop-helix transcription factor; bHLHa17; Class A basic helix-loop-helix protein 17; HGNC:11556; LOW QUALITY PROTEIN: T-cell acute lymphocytic leukemia protein 1; Scl; scl/tal1; SCL/tal-1; SCL-alpha; SCL-beta; si:ch211-135j1.3; stem cell leukemia protein; stem cell leukemia protein SCL; stem cell protein; stem-cell leukemia hematopoietic transcription factor; T cell acute lymphocytic leukemia 1; TAL bHLH transcription factor 1, erythroid differentiation factor; TAL1; Tal-1; tal-1 product; T-cell acute lymphocytic leukemia 1; T-cell acute lymphocytic leukemia protein 1; T-cell acute lymphocytic leukemia protein 1 homolog; T-cell leukemia/lymphoma protein 5; TCL5; transcription factor; zgc:85605; zSCL
Host Species Mouse
Immunogen Full length human recombinant protein of human TAL1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6886
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.