Learn More
TAL1 Mouse anti-Human, Clone: OTI4A6, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF503896

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
TAL-1 is a basic helix-loop-helix transcription. It regulates differentiation and survival during hemopoiesis. TAL-1 is also implicated in the genesis of hemopoietic maligcies and it may play an important role in hemopoietic differentiation. It serves as a positive regulator of eryhtroid differentiation. Mutations are associated with T-cell leukemia and melanoma. TAL-1 is know to bind to the LIM domain containing protein Rhombotin-2.Specifications
TAL1 | |
Monoclonal | |
Unconjugated | |
TAL1 | |
SCL, TCL5, bHLHa17, tal-1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
6886 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI4A6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P17542 | |
TAL1 | |
Full length human recombit protein of human TAL1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.