Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TAL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541498
Description
Peptide sequence: TRKIFTNTRE RWRQQNVNSA FAKLRKLIPT HPPDKKLSKN ETLRLAMRYI Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 92%; Rabbit: 92%; Rat: 93%; Zebrafish: 92%.
TAL2 is a helix-loop-helix protein. Translocations between this gene on chromosome 9 and the T-cell receptor beta-chain locus on chromosome 7 have been associated with activation of the T-cell acute lymphocytic leukemia 2 gene and T-cell acute lymphoblastic leukemia.
Specifications
TAL2 | |
Polyclonal | |
Unconjugated | |
Tal2 | |
bHLHa19; Class A basic helix-loop-helix protein 19; T cell acute lymphocytic leukemia 2; tail anomaly lethal 2; Tail2; TAL bHLH transcription factor 2; Tal2; TAL-2; T-cell acute lymphocytic leukemia 2; T-cell acute lymphocytic leukemia protein 2; T-cell acute lymphocytic leukemia protein 2 homolog; zgc:111884 | |
Rabbit | |
Affinity chromatography | |
RUO | |
6887 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q16559 | |
Tal2 | |
Synthetic peptide directed towards the N-terminal of human TAL2 (aa 2-51). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction