Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Talampanel
GREENER_CHOICESDP

Catalog No. 503210R Shop All R&D Systems Products
Change view
Click to view available options
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
503210R 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 503210R Supplier Tocris Bioscience™ Supplier No. 5032/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Non-competitive AMPA/kainate antagonist

Talampanel is a non-competitive AMPA/kainate receptor antagonist that displays 2.3-3-fold more potent activity than GYKI 52466 (Cat. No. 1454). Potentiates the anticonvulsive activity of antiepileptic drugs in animal models of seizures. Orally active.

Specifications

Quantity 10 mg
Product Type Talampanel
Purity >98%
Molecular Weight (g/mol) 337.37
Inhibitors Talampanel

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.