Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Talc
SCI_ED

Catalog No. s25799 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
Packaging:
Solid
This item is not returnable. View return policy
This item is not returnable. View return policy

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 14807-96-6
Molecular Formula H2Mg3O12Si4
Molecular Weight (g/mol) 379.26
MDL Number MFCD00792903
InChI Key KNXVOGGZOFOROK-UHFFFAOYSA-N
Synonym magnesium silicate, hydrous, talc powder, talc, cp, talc,-350 mesh, talc, powder, 10 mum, 3mgo.4o2si.h2o, talc, vetec tm reagent grade, talc, tested according to ph.eur., magnesium silicate monohydrate talc, tris @magnesium oxide tetrakis silica hydrate
PubChem CID 16211421
IUPAC Name trimagnesium(2+) bis(oxosilanebis(olate)) dihydroxysilanoylolate
SMILES [Mg++].[Mg++].[Mg++].O[Si]([O-])=O.O[Si]([O-])=O.[O-][Si]([O-])=O.[O-][Si]([O-])=O

Specifications

Quantity 500 g
Assay Percent Range >99% w/v
Linear Formula 3MgO·4SiO2·H2O
Formula Weight 166.4
Packaging Solid
Density 2.5 to 2.8g/mL
Chemical Name or Material Talc
ShelfLife 5 years
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.