Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Talc, 95% Powder, 325 Mesh, Lab Grade, Innovating Science™
SCI_ED

Catalog No. S02277 Shop All Aldon Products
Click to view available options
Quantity:
500 g
WARNING! This product can expose you to a chemical, crystalline silica, which is known to the State of California to cause cancer.
WARNING! This product can expose you to a chemical, crystalline silica, which is known to the State of California to cause cancer.

Chemical Identifiers

CAS 14807-96-6

Specifications

Physical Form Flakes or Powder
Quantity 500 g
Grade Laboratory
Concentration or Composition (by Analyte or Components) 95%
Packaging HDPE
Chemical Name or Material Talc
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.