Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Talc (Powder/Laboratory), Fisher Chemical™

Catalog No. p-34481 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle

Specifications

Chemical Name or Material Talc
Melting Point 800°C
Color White
CAS 14807-96-6, 14808-60-7
Physical Form Powder/Solid
Quantity 500 g
Molecular Formula 3MgO4SiO2H2O
MDL Number MFCD00084820
Synonym Soapstone, Talcum
Grade Laboratory
Identification Pass Test
Packaging Poly Bottle
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.